Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606670 MIM: 607524 MIM: 607525 MIM: 615714 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PPP1R11 - protein phosphatase 1 regulatory inhibitor subunit 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF39 - ring finger protein 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNRD1 - zinc ribbon domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278785.1 | Intron | NP_001265714.1 | ||||
NM_001278786.1 | Intron | NP_001265715.1 | ||||
NM_014596.5 | Intron | NP_055411.1 | ||||
NM_170783.3 | Intron | NP_740753.1 |
ZNRD1ASP - zinc ribbon domain containing 1 antisense, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |