Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602597 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL9 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BCL9 - B-cell CLL/lymphoma 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004326.3 | Intron | NP_004317.2 | ||||
XM_005272971.4 | Intron | XP_005273028.1 | ||||
XM_005277417.3 | Intron | XP_005277474.1 | ||||
XM_005277418.3 | Intron | XP_005277475.1 | ||||
XM_017002026.1 | Intron | XP_016857515.1 |