Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCACGCTGGCCCCTTAAACTTCC[G/T]TAACCATGTCAGGCACATGCTTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610340 MIM: 604574 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPB41L4B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EPB41L4B - erythrocyte membrane protein band 4.1 like 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018424.3 | Intron | NP_060894.2 | ||||
NM_019114.4 | Intron | NP_061987.3 | ||||
XM_011518790.1 | Intron | XP_011517092.1 | ||||
XM_011518791.2 | Intron | XP_011517093.1 | ||||
XM_011518792.2 | Intron | XP_011517094.1 | ||||
XM_011518794.2 | Intron | XP_011517096.1 | ||||
XM_017014813.1 | Intron | XP_016870302.1 | ||||
XM_017014814.1 | Intron | XP_016870303.1 | ||||
XM_017014815.1 | Intron | XP_016870304.1 |
FRRS1L - ferric chelate reductase 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |