Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTGCAGACAGCCGCCTTCGTGC[C/T]GCGTCCTCACATGGTGGAAAAAGAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614780 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNX10 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SNX10 - sorting nexin 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199835.1 | Intron | NP_001186764.1 | ||||
NM_001199837.1 | Intron | NP_001186766.1 | ||||
NM_001199838.1 | Intron | NP_001186767.1 | ||||
NM_001318198.1 | Intron | NP_001305127.1 | ||||
NM_001318199.1 | Intron | NP_001305128.1 | ||||
NM_013322.2 | Intron | NP_037454.2 | ||||
XM_006715712.2 | Intron | XP_006715775.1 | ||||
XM_017012085.1 | Intron | XP_016867574.1 | ||||
XM_017012086.1 | Intron | XP_016867575.1 | ||||
XM_017012087.1 | Intron | XP_016867576.1 | ||||
XM_017012088.1 | Intron | XP_016867577.1 |