Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153370 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGAL PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ITGAL - integrin subunit alpha L | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF747 - zinc finger protein 747 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305018.1 | 2638 | UTR 3 | NP_001291947.1 | |||
NM_001305019.1 | 2638 | UTR 3 | NP_001291948.1 | |||
NM_001305020.1 | 2638 | UTR 3 | NP_001291949.1 | |||
NM_023931.3 | 2638 | UTR 3 | NP_076420.1 |
ZNF768 - zinc finger protein 768 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024671.3 | 2638 | Intron | NP_078947.3 | |||
XM_017023665.1 | 2638 | Intron | XP_016879154.1 | |||
XM_017023666.1 | 2638 | Intron | XP_016879155.1 |
Set Membership: |
HapMap |