Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TYW1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TYW1B - tRNA-yW synthesizing protein 1 homolog B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145440.2 | Intron | NP_001138912.2 | ||||
XM_011516230.2 | Intron | XP_011514532.1 | ||||
XM_011516231.2 | Intron | XP_011514533.1 | ||||
XM_011516232.2 | Intron | XP_011514534.1 | ||||
XM_011516233.2 | Intron | XP_011514535.1 | ||||
XM_017012221.1 | Intron | XP_016867710.1 | ||||
XM_017012222.1 | Intron | XP_016867711.1 |
Set Membership: |
HapMap |