Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACTGAAGTCTTGAACCTGTCGGT[C/G]AAGGTTGGAACCAACTTCTTCTAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615130 | ||||||||||||||||||||
Literature Links: |
GALNT11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GALNT11 - polypeptide N-acetylgalactosaminyltransferase 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304514.1 | Intron | NP_001291443.1 | ||||
NM_022087.3 | Intron | NP_071370.2 | ||||
XM_006716082.2 | Intron | XP_006716145.1 | ||||
XM_006716083.2 | Intron | XP_006716146.1 | ||||
XM_006716084.2 | Intron | XP_006716147.1 | ||||
XM_017012499.1 | Intron | XP_016867988.1 | ||||
XM_017012500.1 | Intron | XP_016867989.1 |