Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGTTCTGGCCTCAAAGCATAAAT[A/G]CCTCCCCATCCTGTGGGAGGTACTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604942 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKBP1A-SDCBP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FKBP1A-SDCBP2 - FKBP1A-SDCBP2 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SDCBP2 - syndecan binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199784.1 | Intron | NP_001186713.1 | ||||
NM_015685.5 | Intron | NP_056500.2 | ||||
NM_080489.4 | Intron | NP_536737.3 |
SNPH - syntaphilin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |