Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602190 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPHA7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EPHA7 - EPH receptor A7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288629.1 | Intron | NP_001275558.1 | ||||
NM_001288630.1 | Intron | NP_001275559.1 | ||||
NM_004440.3 | Intron | NP_004431.1 | ||||
XM_005248669.2 | Intron | XP_005248726.1 | ||||
XM_005248671.2 | Intron | XP_005248728.1 | ||||
XM_017010365.1 | Intron | XP_016865854.1 | ||||
XM_017010366.1 | Intron | XP_016865855.1 |
Set Membership: |
HapMap |