Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTTTAACCTGGGGTTTTCTGGTT[C/T]TGGGGTTTATATATTTTGGCTGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600879 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NRF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NRF1 - nuclear respiratory factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040110.1 | Intron | NP_001035199.1 | ||||
NM_001293163.1 | Intron | NP_001280092.1 | ||||
NM_001293164.1 | Intron | NP_001280093.1 | ||||
NM_005011.4 | Intron | NP_005002.3 |
Set Membership: |
HapMap |