Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACTGCGCCCAGCCATGGGCACA[C/T]ATTTCATGTGGATAAAGTTGAGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612684 MIM: 605713 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ISM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ISM2 - isthmin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPTLC2 - serine palmitoyltransferase long chain base subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004863.3 | 6522 | UTR 3 | NP_004854.1 | |||
XM_011537384.2 | 6522 | Intron | XP_011535686.1 |
Set Membership: |
HapMap |