Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCCTAAACAACAGAGCTGTCCCG[A/G]CCAAACAGACACAGCTGAAGGCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 243305 MIM: 616717 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INVS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
INVS - inversin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318381.1 | Intron | NP_001305310.1 | ||||
NM_001318382.1 | Intron | NP_001305311.1 | ||||
NM_014425.4 | Intron | NP_055240.2 |
TEX10 - testis expressed 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161584.1 | Intron | NP_001155056.1 | ||||
NM_017746.3 | Intron | NP_060216.2 | ||||
XM_011518798.1 | Intron | XP_011517100.1 | ||||
XM_017014848.1 | Intron | XP_016870337.1 |