Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 189969 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF2F2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GTF2F2 - general transcription factor IIF subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004128.2 | 348 | Intron | NP_004119.1 | |||
XM_011535052.2 | 348 | Intron | XP_011533354.1 | |||
XM_011535053.2 | 348 | Intron | XP_011533355.1 | |||
XM_017020550.1 | 348 | Intron | XP_016876039.1 | |||
XM_017020551.1 | 348 | Intron | XP_016876040.1 |
KCTD4 - potassium channel tetramerization domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198404.2 | 348 | UTR 5 | NP_940686.2 |
Set Membership: |
HapMap |