Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 617019 | |||||||||||||||||||||||
Literature Links: |
TMEM230 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TMEM230 - transmembrane protein 230 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009923.1 | Intron | NP_001009923.1 | ||||
NM_001009924.1 | Intron | NP_001009924.1 | ||||
NM_001009925.1 | Intron | NP_001009925.1 | ||||
NM_014145.4 | Intron | NP_054864.3 | ||||
XM_005260713.3 | Intron | XP_005260770.1 | ||||
XM_011529227.2 | Intron | XP_011527529.1 | ||||
XM_011529229.2 | Intron | XP_011527531.1 | ||||
XM_017027834.1 | Intron | XP_016883323.1 | ||||
XM_017027835.1 | Intron | XP_016883324.1 | ||||
XM_017027836.1 | Intron | XP_016883325.1 |