Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCTGTTTCCTTTGCTGTAACTGC[G/T]CAGATTCCTTAAACTCTGTATCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606058 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAPGEF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RAPGEF4 - Rap guanine nucleotide exchange factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100397.1 | Intron | NP_001093867.1 | ||||
NM_001282899.1 | Intron | NP_001269828.1 | ||||
NM_001282900.1 | Intron | NP_001269829.1 | ||||
NM_001282901.1 | Intron | NP_001269830.1 | ||||
NM_007023.3 | Intron | NP_008954.2 | ||||
XM_005246246.2 | Intron | XP_005246303.1 | ||||
XM_006712205.3 | Intron | XP_006712268.1 | ||||
XM_011510505.2 | Intron | XP_011508807.1 | ||||
XM_017003196.1 | Intron | XP_016858685.1 | ||||
XM_017003197.1 | Intron | XP_016858686.1 |
RAPGEF4-AS1 - RAPGEF4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |