Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTCCACTTTTTACTGTCATCAT[G/T]TACCAGCAATTTGAATCAACTTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105372493 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105372493 - uncharacterized LOC105372493 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMF1 - proteasome inhibitor subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323407.1 | Intron | NP_001310336.1 | ||||
NM_001323408.1 | Intron | NP_001310337.1 | ||||
NM_001323409.1 | Intron | NP_001310338.1 | ||||
NM_001323410.1 | Intron | NP_001310339.1 | ||||
NM_006814.4 | Intron | NP_006805.2 | ||||
NM_178578.3 | Intron | NP_848693.2 |
Set Membership: |
HapMap |