Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600715 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MTX3 - metaxin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THBS4 - thrombospondin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306212.1 | Intron | NP_001293141.1 | ||||
NM_001306213.1 | Intron | NP_001293142.1 | ||||
NM_001306214.1 | Intron | NP_001293143.1 | ||||
NM_003248.5 | Intron | NP_003239.2 | ||||
XM_017009798.1 | Intron | XP_016865287.1 | ||||
XM_017009799.1 | Intron | XP_016865288.1 | ||||
XM_017009800.1 | Intron | XP_016865289.1 |