Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTATTGGCTTCATATTACTTTTT[A/G]AAATGTTTTATTTTTGTAAAAGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607843 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PKHD1L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PKHD1L1 - polycystic kidney and hepatic disease 1 (autosomal recessive)-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177531.4 | Intron | NP_803875.2 | ||||
XM_011517371.2 | Intron | XP_011515673.1 | ||||
XM_017013969.1 | Intron | XP_016869458.1 | ||||
XM_017013970.1 | Intron | XP_016869459.1 | ||||
XM_017013971.1 | Intron | XP_016869460.1 | ||||
XM_017013972.1 | Intron | XP_016869461.1 | ||||
XM_017013973.1 | Intron | XP_016869462.1 | ||||
XM_017013974.1 | Intron | XP_016869463.1 |