Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGTGACTGGCTGGAGAAATAAG[G/T]TAGGGAGAATCTAGATATGGTTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608472 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ST6GAL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ST6GAL2 - ST6 beta-galactoside alpha-2,6-sialyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142351.1 | 4832 | UTR 3 | NP_001135823.1 | |||
NM_001142352.1 | 4832 | Intron | NP_001135824.1 | |||
NM_001322362.1 | 4832 | UTR 3 | NP_001309291.1 | |||
NM_032528.2 | 4832 | UTR 3 | NP_115917.1 | |||
XM_006712802.1 | 4832 | UTR 3 | XP_006712865.1 | |||
XM_011511999.2 | 4832 | UTR 3 | XP_011510301.1 | |||
XM_011512000.2 | 4832 | Intron | XP_011510302.1 | |||
XM_011512001.2 | 4832 | Intron | XP_011510303.1 | |||
XM_017005110.1 | 4832 | Intron | XP_016860599.1 |
Set Membership: |
HapMap |