Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TTBK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TTBK1 - tau tubulin kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011514947.2 | Intron | XP_011513249.1 | ||||
XM_011514948.2 | Intron | XP_011513250.1 | ||||
XM_017011364.1 | Intron | XP_016866853.1 | ||||
XM_017011365.1 | Intron | XP_016866854.1 | ||||
XM_017011366.1 | Intron | XP_016866855.1 | ||||
XM_017011367.1 | Intron | XP_016866856.1 | ||||
XM_017011368.1 | Intron | XP_016866857.1 |