Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGTATGCTGAGATTATAATAAAC[A/G]CAGAATTCTGATTCAGTATTTCTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 133440 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF4E PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EIF4E - eukaryotic translation initiation factor 4E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130678.1 | 4046 | UTR 3 | NP_001124150.1 | |||
NM_001130679.1 | 4046 | UTR 3 | NP_001124151.1 | |||
NM_001968.3 | 4046 | UTR 3 | NP_001959.1 | |||
XM_006714126.3 | 4046 | Intron | XP_006714189.1 |
Set Membership: |
HapMap |