Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTATTTCTGGGTAATTCATATCCT[C/T]GCTATAGCCTAGATCTAAAAATGAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP43 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CFAP43 - cilia and flagella associated protein 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025145.5 | Intron | NP_079421.5 | ||||
XM_005270171.2 | Intron | XP_005270228.1 | ||||
XM_005270172.3 | Intron | XP_005270229.1 | ||||
XM_011540196.2 | Intron | XP_011538498.1 | ||||
XM_011540197.2 | Intron | XP_011538499.1 | ||||
XM_011540198.2 | Intron | XP_011538500.1 | ||||
XM_011540199.2 | Intron | XP_011538501.1 | ||||
XM_011540200.2 | Intron | XP_011538502.1 | ||||
XM_011540201.2 | Intron | XP_011538503.1 | ||||
XM_011540202.2 | Intron | XP_011538504.1 | ||||
XM_011540203.2 | Intron | XP_011538505.1 | ||||
XM_017016681.1 | Intron | XP_016872170.1 | ||||
XM_017016682.1 | Intron | XP_016872171.1 | ||||
XM_017016683.1 | Intron | XP_016872172.1 | ||||
XM_017016684.1 | Intron | XP_016872173.1 |