Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615932 | ||||||||||||||||||||
Literature Links: |
KCTD20 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCTD20 - potassium channel tetramerization domain containing 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286579.1 | Intron | NP_001273508.1 | ||||
NM_001286580.1 | Intron | NP_001273509.1 | ||||
NM_173562.4 | Intron | NP_775833.2 | ||||
XM_005248940.4 | Intron | XP_005248997.1 | ||||
XM_006715023.2 | Intron | XP_006715086.1 | ||||
XM_011514398.2 | Intron | XP_011512700.1 |
PXT1 - peroxisomal, testis specific 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |