Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RALGAPA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RALGAPA2 - Ral GTPase activating protein catalytic alpha subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020343.3 | 6431 | UTR 3 | NP_065076.2 | |||
XM_005260768.2 | 6431 | Intron | XP_005260825.1 | |||
XM_006723598.1 | 6431 | UTR 3 | XP_006723661.1 | |||
XM_006723599.1 | 6431 | UTR 3 | XP_006723662.1 | |||
XM_011529309.1 | 6431 | Intron | XP_011527611.1 | |||
XM_011529310.2 | 6431 | UTR 3 | XP_011527612.1 | |||
XM_011529311.2 | 6431 | UTR 3 | XP_011527613.2 | |||
XM_017027972.1 | 6431 | Intron | XP_016883461.1 | |||
XM_017027973.1 | 6431 | UTR 3 | XP_016883462.1 | |||
XM_017027974.1 | 6431 | UTR 3 | XP_016883463.1 | |||
XM_017027975.1 | 6431 | UTR 3 | XP_016883464.1 | |||
XM_017027976.1 | 6431 | UTR 3 | XP_016883465.1 | |||
XM_017027977.1 | 6431 | Intron | XP_016883466.1 |