Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGGCAAATGCAACCGTCATTTTT[A/C]GAGTCTTCAGCCGTGCTCTTGGTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138850 | ||||||||||||||||||||
Literature Links: |
GNRHR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GNRHR - gonadotropin releasing hormone receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000406.2 | 2548 | Missense Mutation | CGA,CTA | R,L 266 | NP_000397.1 | |
NM_001012763.1 | 2548 | Silent Mutation | TCG,TCT | S,S 223 | NP_001012781.1 |