Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAAAGAATAGGACTTTTCCTCTA[C/T]CTTCAAACCACTTAGGAAGAATACC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613946 MIM: 602878 MIM: 606474 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC5G PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DNAJC5G - DnaJ heat shock protein family (Hsp40) member C5 gamma | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC30A3 - solute carrier family 30 member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM54 - tripartite motif containing 54 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032546.3 | Intron | NP_115935.3 | ||||
NM_187841.2 | Intron | NP_912730.2 |
Set Membership: |
HapMap |