Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAGGAGTTTCAGTGTGATACTGA[C/T]GAGGACCAAATTTTGATACATGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf57 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C7orf57 - chromosome 7 open reading frame 57 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100159.2 | Intron | NP_001093629.1 | ||||
NM_001267865.1 | Intron | NP_001254794.1 | ||||
NM_001267866.1 | Intron | NP_001254795.1 | ||||
XM_006715649.3 | Intron | XP_006715712.1 | ||||
XM_006715650.2 | Intron | XP_006715713.1 | ||||
XM_011515121.1 | Intron | XP_011513423.1 | ||||
XM_011515122.2 | Intron | XP_011513424.1 | ||||
XM_011515123.2 | Intron | XP_011513425.1 |
SUN3 - Sad1 and UNC84 domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |