Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGAGGCCTGCTGCCTCCCTGATG[C/T]CTGTCCATAAAGCAGGGTGGGAGAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614950 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM17 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TMEM17 - transmembrane protein 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198276.2 | 1332 | Intron | NP_938017.2 | |||
XM_011532693.2 | 1332 | Intron | XP_011530995.1 | |||
XM_011532694.2 | 1332 | UTR 3 | XP_011530996.1 | |||
XM_017003575.1 | 1332 | Intron | XP_016859064.1 |