Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600409 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPARD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PPARD - peroxisome proliferator activated receptor delta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171818.1 | Intron | NP_001165289.1 | ||||
NM_001171819.1 | Intron | NP_001165290.1 | ||||
NM_001171820.1 | Intron | NP_001165291.1 | ||||
NM_006238.4 | Intron | NP_006229.1 | ||||
NM_177435.2 | Intron | NP_803184.1 | ||||
XM_005249193.1 | Intron | XP_005249250.1 | ||||
XM_006715120.1 | Intron | XP_006715183.1 | ||||
XM_006715123.1 | Intron | XP_006715186.1 | ||||
XM_011514707.1 | Intron | XP_011513009.1 | ||||
XM_011514709.1 | Intron | XP_011513011.1 | ||||
XM_011514710.1 | Intron | XP_011513012.1 | ||||
XM_017010971.1 | Intron | XP_016866460.1 | ||||
XM_017010972.1 | Intron | XP_016866461.1 | ||||
XM_017010973.1 | Intron | XP_016866462.1 | ||||
XM_017010974.1 | Intron | XP_016866463.1 | ||||
XM_017010975.1 | Intron | XP_016866464.1 |
Set Membership: |
HapMap |