Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614818 | ||||||||||||||||||||
Literature Links: |
FRY PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FRY - FRY microtubule binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023037.2 | Intron | NP_075463.2 | ||||
XM_006719749.3 | Intron | XP_006719812.1 | ||||
XM_017020303.1 | Intron | XP_016875792.1 | ||||
XM_017020304.1 | Intron | XP_016875793.1 | ||||
XM_017020305.1 | Intron | XP_016875794.1 | ||||
XM_017020306.1 | Intron | XP_016875795.1 | ||||
XM_017020307.1 | Intron | XP_016875796.1 |
FRY-AS1 - FRY antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |