Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604086 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF155 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF155 - zinc finger protein 155 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001260486.1 | 2532 | Intron | NP_001247415.1 | |||
NM_001260487.1 | 2532 | Intron | NP_001247416.1 | |||
NM_001260488.1 | 2532 | Intron | NP_001247417.1 | |||
NM_003445.3 | 2532 | Intron | NP_003436.3 | |||
NM_198089.2 | 2532 | Intron | NP_932355.2 | |||
XM_005259215.3 | 2532 | UTR 5 | XP_005259272.1 | |||
XM_011527278.2 | 2532 | Intron | XP_011525580.1 | |||
XM_011527279.2 | 2532 | UTR 5 | XP_011525581.1 | |||
XM_017027248.1 | 2532 | Intron | XP_016882737.1 |
ZNF221 - zinc finger protein 221 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297588.1 | 2532 | Intron | NP_001284517.1 | |||
NM_001297589.1 | 2532 | Intron | NP_001284518.1 | |||
NM_013359.2 | 2532 | Intron | NP_037491.2 | |||
XM_017027232.1 | 2532 | Intron | XP_016882721.1 | |||
XM_017027233.1 | 2532 | UTR 3 | XP_016882722.1 |
Set Membership: |
HapMap |