Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608427 MIM: 602544 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PACRG PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PACRG - PARK2 coregulated | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PARK2 - parkin RBR E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004562.2 | Intron | NP_004553.2 | ||||
NM_013987.2 | Intron | NP_054642.2 | ||||
NM_013988.2 | Intron | NP_054643.2 | ||||
XM_011535863.1 | Intron | XP_011534165.1 | ||||
XM_017010908.1 | Intron | XP_016866397.1 | ||||
XM_017010909.1 | Intron | XP_016866398.1 |