Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACTTTACTGCATGTAACAGAGAC[A/G]ACTCCCACCTATCAGTATCTTAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600111 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC1A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC1A3 - solute carrier family 1 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166695.2 | Intron | NP_001160167.1 | ||||
NM_001166696.2 | Intron | NP_001160168.1 | ||||
NM_001289939.1 | Intron | NP_001276868.1 | ||||
NM_001289940.1 | Intron | NP_001276869.1 | ||||
NM_004172.4 | Intron | NP_004163.3 | ||||
XM_005248342.2 | Intron | XP_005248399.1 | ||||
XM_011514084.2 | Intron | XP_011512386.1 |
Set Membership: |
HapMap |