Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTTTTGTTACTCCAGTTGGTGGT[G/T]AAGAGGCCAGGCTGAGGCAGAGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PPP1R36 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP1R36 - protein phosphatase 1 regulatory subunit 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172365.2 | Intron | NP_758953.1 | ||||
XM_005267354.3 | Intron | XP_005267411.1 | ||||
XM_005267355.3 | Intron | XP_005267412.1 | ||||
XM_011536468.2 | Intron | XP_011534770.1 |