Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGATAGGTGATCGTGCACCAGCT[A/G]GAAGTAAGAGGACCCTGTGAGCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603053 | ||||||||||||||||||||
Literature Links: |
HAT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HAT1 - histone acetyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003642.3 | Intron | NP_003633.1 | ||||
XM_006712808.3 | Intron | XP_006712871.1 | ||||
XM_011512027.2 | Intron | XP_011510329.1 | ||||
XM_017005136.1 | Intron | XP_016860625.1 | ||||
XM_017005137.1 | Intron | XP_016860626.1 |