Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAAGACATTACGCTTAAACTCAC[C/T]GTCTGCCAGCAAGTCTCCTAGACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
61 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERICH3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERICH3 - glutamate rich 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002912.4 | 4830 | UTR 3 | NP_001002912.4 | |||
XM_017000274.1 | 4830 | UTR 3 | XP_016855763.1 | |||
XM_017000275.1 | 4830 | Intron | XP_016855764.1 | |||
XM_017000276.1 | 4830 | UTR 3 | XP_016855765.1 | |||
XM_017000277.1 | 4830 | UTR 3 | XP_016855766.1 | |||
XM_017000278.1 | 4830 | UTR 3 | XP_016855767.1 | |||
XM_017000279.1 | 4830 | UTR 3 | XP_016855768.1 | |||
XM_017000280.1 | 4830 | UTR 3 | XP_016855769.1 | |||
XM_017000281.1 | 4830 | UTR 3 | XP_016855770.1 | |||
XM_017000282.1 | 4830 | UTR 3 | XP_016855771.1 | |||
XM_017000283.1 | 4830 | UTR 3 | XP_016855772.1 | |||
XM_017000284.1 | 4830 | UTR 3 | XP_016855773.1 | |||
XM_017000285.1 | 4830 | UTR 3 | XP_016855774.1 |
ERICH3-AS1 - ERICH3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |