Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606915 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR63 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GPR63 - G protein-coupled receptor 63 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143957.2 | 2678 | UTR 3 | NP_001137429.1 | |||
NM_030784.3 | 2678 | UTR 3 | NP_110411.1 | |||
XM_006715570.3 | 2678 | UTR 3 | XP_006715633.1 | |||
XM_011536153.2 | 2678 | UTR 3 | XP_011534455.1 | |||
XM_011536154.2 | 2678 | UTR 3 | XP_011534456.1 | |||
XM_011536155.2 | 2678 | UTR 3 | XP_011534457.1 | |||
XM_011536157.2 | 2678 | UTR 3 | XP_011534459.1 | |||
XM_011536159.2 | 2678 | UTR 3 | XP_011534461.1 | |||
XM_017011334.1 | 2678 | UTR 3 | XP_016866823.1 |
Set Membership: |
HapMap |