Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCATTAGTGAAATTTTAGGCCTGA[C/T]ATGATGGCTCACGCTTGTAATCCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610694 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PGPEP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PGPEP1 - pyroglutamyl-peptidase I | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |