Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGATTTAGCTTTGTAGAGATGTTC[C/T]GCTAATATCCATAACTAATCTGAGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 264900 MIM: 229000 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
F11 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
F11 - coagulation factor XI | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000128.3 | Intron | NP_000119.1 | ||||
XM_005262821.3 | Intron | XP_005262878.1 | ||||
XM_005262822.3 | Intron | XP_005262879.1 | ||||
XM_005262823.3 | Intron | XP_005262880.1 | ||||
XM_006714137.2 | Intron | XP_006714200.1 | ||||
XM_017007884.1 | Intron | XP_016863373.1 | ||||
XM_017007885.1 | Intron | XP_016863374.1 | ||||
XM_017007886.1 | Intron | XP_016863375.1 |
KLKB1 - kallikrein B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |