Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
POLR3E PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POLR3E - RNA polymerase III subunit E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258033.1 | Intron | NP_001244962.1 | ||||
NM_001258034.1 | Intron | NP_001244963.1 | ||||
NM_001258035.1 | Intron | NP_001244964.1 | ||||
NM_001258036.1 | Intron | NP_001244965.1 | ||||
NM_018119.3 | Intron | NP_060589.1 | ||||
XM_006721063.2 | Intron | XP_006721126.1 | ||||
XM_011545892.1 | Intron | XP_011544194.1 | ||||
XM_011545893.2 | Intron | XP_011544195.1 | ||||
XM_017023443.1 | Intron | XP_016878932.1 |