Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
ZBBX PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ZBBX - zinc finger B-box domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199201.1 | Intron | NP_001186130.1 | ||||
NM_001199202.1 | Intron | NP_001186131.1 | ||||
NM_024687.3 | Intron | NP_078963.2 | ||||
XM_011513144.1 | Intron | XP_011511446.1 | ||||
XM_011513145.2 | Intron | XP_011511447.2 | ||||
XM_011513148.2 | Intron | XP_011511450.2 | ||||
XM_011513149.2 | Intron | XP_011511451.2 | ||||
XM_011513151.2 | Intron | XP_011511453.2 | ||||
XM_011513152.2 | Intron | XP_011511454.2 | ||||
XM_011513153.2 | Intron | XP_011511455.2 | ||||
XM_011513155.1 | Intron | XP_011511457.1 | ||||
XM_011513156.1 | Intron | XP_011511458.1 | ||||
XM_017007186.1 | Intron | XP_016862675.1 | ||||
XM_017007187.1 | Intron | XP_016862676.1 | ||||
XM_017007188.1 | Intron | XP_016862677.1 | ||||
XM_017007189.1 | Intron | XP_016862678.1 | ||||
XM_017007190.1 | Intron | XP_016862679.1 |