Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAATTTAGATAAATATTAATCAG[C/T]ATGGGAATGGTTAAGTAAACTACAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615655 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZRANB3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZRANB3 - zinc finger RANBP2-type containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286568.1 | Intron | NP_001273497.1 | ||||
NM_001286569.1 | Intron | NP_001273498.1 | ||||
NM_032143.3 | Intron | NP_115519.2 | ||||
XM_005263809.1 | Intron | XP_005263866.1 | ||||
XM_006712788.1 | Intron | XP_006712851.1 | ||||
XM_011511958.2 | Intron | XP_011510260.1 | ||||
XM_011511959.2 | Intron | XP_011510261.2 | ||||
XM_011511960.1 | Intron | XP_011510262.1 | ||||
XM_011511961.2 | Intron | XP_011510263.2 | ||||
XM_011511963.1 | Intron | XP_011510265.1 | ||||
XM_011511966.2 | Intron | XP_011510268.2 | ||||
XM_017005065.1 | Intron | XP_016860554.1 | ||||
XM_017005066.1 | Intron | XP_016860555.1 | ||||
XM_017005067.1 | Intron | XP_016860556.1 | ||||
XM_017005068.1 | Intron | XP_016860557.1 | ||||
XM_017005069.1 | Intron | XP_016860558.1 | ||||
XM_017005070.1 | Intron | XP_016860559.1 | ||||
XM_017005071.1 | Intron | XP_016860560.1 |
Set Membership: |
HapMap |