Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGGGAGAGGACAACTAGAAACC[A/G]CACATGGACTTCTCCTGGGCTCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610531 | ||||||||||||||||||||
Literature Links: |
FAM126A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM126A - family with sequence similarity 126 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032581.3 | Intron | NP_115970.2 | ||||
XM_005249894.3 | Intron | XP_005249951.1 | ||||
XM_005249895.2 | Intron | XP_005249952.1 | ||||
XM_011515589.2 | Intron | XP_011513891.1 | ||||
XM_011515590.1 | Intron | XP_011513892.1 |