Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCGGGACTGGCTGTACTTGCACCG[C/T]TACAACGCCTACCCCTCAGAGCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607294 | ||||||||||||||||||||
Literature Links: |
DLGAP4-AS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DLGAP4-AS1 - DLGAP4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TGIF2 - TGFB induced factor homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199513.1 | 239 | Silent Mutation | CGC,CGT | R,R 41 | NP_001186442.1 | |
NM_001199514.1 | 239 | Silent Mutation | CGC,CGT | R,R 41 | NP_001186443.1 | |
NM_001199515.1 | 239 | Silent Mutation | CGC,CGT | R,R 41 | NP_001186444.1 | |
NM_021809.6 | 239 | Silent Mutation | CGC,CGT | R,R 41 | NP_068581.1 |
TGIF2-C20orf24 - TGIF2-C20orf24 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199535.1 | 239 | Silent Mutation | CGC,CGT | R,R 41 | NP_001186464.1 |