Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116949 MIM: 117140 MIM: 610674 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC25B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDC25B - cell division cycle 25B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287516.1 | Intron | NP_001274445.1 | ||||
NM_001287517.1 | Intron | NP_001274446.1 | ||||
NM_001287518.1 | Intron | NP_001274447.1 | ||||
NM_001287519.1 | Intron | NP_001274448.1 | ||||
NM_001287520.1 | Intron | NP_001274449.1 | ||||
NM_001287522.1 | Intron | NP_001274451.1 | ||||
NM_001287524.1 | Intron | NP_001274453.1 | ||||
NM_004358.4 | Intron | NP_004349.1 | ||||
NM_021872.3 | Intron | NP_068658.1 | ||||
NM_021873.3 | Intron | NP_068659.1 |
CENPB - centromere protein B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPEF1 - sperm flagellar 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |