Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606145 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC2A10 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC2A10 - solute carrier family 2 member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030777.3 | Intron | NP_110404.1 | ||||
XM_011529060.2 | Intron | XP_011527362.1 | ||||
XM_011529061.2 | Intron | XP_011527363.1 | ||||
XM_011529062.2 | Intron | XP_011527364.1 | ||||
XM_011529063.2 | Intron | XP_011527365.1 | ||||
XM_011529064.2 | Intron | XP_011527366.1 | ||||
XM_011529065.2 | Intron | XP_011527367.1 | ||||
XM_017028087.1 | Intron | XP_016883576.1 |