Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118504 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRNA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CHRNA4 - cholinergic receptor nicotinic alpha 4 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000744.6 | 4827 | UTR 3 | NP_000735.1 | |||
NM_001256573.1 | 4827 | UTR 3 | NP_001243502.1 | |||
XM_011528524.1 | 4827 | Intron | XP_011526826.1 | |||
XM_017027624.1 | 4827 | Intron | XP_016883113.1 | |||
XM_017027625.1 | 4827 | Intron | XP_016883114.1 |
COL20A1 - collagen type XX alpha 1 chain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |