Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604871 | ||||||||||||||||||||
Literature Links: |
MMP24 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MMP24 - matrix metallopeptidase 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006690.3 | Intron | NP_006681.1 | ||||
XM_011528500.2 | Intron | XP_011526802.1 | ||||
XM_017027597.1 | Intron | XP_016883086.1 | ||||
XM_017027598.1 | Intron | XP_016883087.1 |
MT1P3 - metallothionein 1 pseudogene 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |