Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATTGAAAGTCATAGCAAGTGCA[A/G]TTTATCACATTATTTATTTCATAAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
44 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164175 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
POU2F1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
POU2F1 - POU class 2 homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198783.1 | Intron | NP_001185712.1 | ||||
NM_001198786.1 | Intron | NP_001185715.1 | ||||
NM_002697.3 | Intron | NP_002688.3 | ||||
XM_011509653.2 | Intron | XP_011507955.1 | ||||
XM_011509654.2 | Intron | XP_011507956.1 | ||||
XM_011509655.1 | Intron | XP_011507957.1 | ||||
XM_017001508.1 | Intron | XP_016856997.1 |