Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300753 | ||||||||||||||||||||
Literature Links: |
APOO PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APOO - apolipoprotein O | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024122.4 | Intron | NP_077027.1 | ||||
XM_011545584.2 | Intron | XP_011543886.1 | ||||
XM_011545585.2 | Intron | XP_011543887.1 | ||||
XM_011545586.2 | Intron | XP_011543888.1 | ||||
XM_011545587.2 | Intron | XP_011543889.1 | ||||
XM_017029837.1 | Intron | XP_016885326.1 |